siRNA Details
| Gene Symbol: ACAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014716.3 | NP_055531.1 | 11 | 1080 | ||
| GenBank mRNA | BC018543.1 | - | 11 | 1055 | 4342840 | |
| BT009788.1 | - | 11 | 874 | |||
| D30758.1 | - | 11 | 1062 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACAP1| Gene Symbol: ACAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014716.3 | NP_055531.1 | 11 | 1080 | ||
| GenBank mRNA | BC018543.1 | - | 11 | 1055 | 4342840 | |
| BT009788.1 | - | 11 | 874 | |||
| D30758.1 | - | 11 | 1062 | |||