siRNA Details
| Gene Symbol: RNF144A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014746.4 | NP_055561.2 | 9 | 1335 | ||
| XM_005246200.3 | XP_005246257.1 | 9 | 2526 | |||
| XM_005246202.4 | XP_005246259.1 | 8 | 1166 | |||
| XM_017005396.1 | XP_016860885.1 | 9 | 2502 | |||
| XM_017005397.1 | XP_016860886.1 | 9 | 1160 | |||
| XM_017005404.1 | XP_016860893.1 | 8 | 1143 | |||
| GenBank mRNA | AK293854.1 | - | 8 | 990 | ||
| BC050373.1 | - | 9 | 1275 | 5722878 | ||
| D79983.1 | - | 9 | 1241 | |||

RNF144A
Genomic Map