siRNA Details
| Gene Symbol: RNF144A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014746.4 | NP_055561.2 | 9 | 1916 | ||
| XM_005246200.3 | XP_005246257.1 | 9 | 3107 | |||
| XM_005246202.4 | XP_005246259.1 | 8 | 1747 | |||
| XM_017005396.1 | XP_016860885.1 | 9 | 3083 | |||
| XM_017005397.1 | XP_016860886.1 | 9 | 1741 | |||
| XM_017005404.1 | XP_016860893.1 | 8 | 1724 | |||
| GenBank mRNA | D79983.1 | - | 9 | 1822 | ||

RNF144A
Genomic Map