siRNA Details
| Gene Symbol: KIAA0586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244189.1 | NP_001231118.1 | 18 | 2672 | ||
| NM_001244190.1 | NP_001231119.1 | 17 | 2515 | |||
| NM_001244191.1 | NP_001231120.1 | 16 | 2323 | |||
| NM_001244192.1 | NP_001231121.1 | 17 | 2446 | |||
| NM_001244193.1 | NP_001231122.1 | 13 | 1959 | |||
| NM_014749.3 | NP_055564.3 | 15 | 2314 | |||
| GenBank mRNA | AB011158.1 | - | 17 | 2498 | ||
| AK302579.1 | - | 13 | 1961 | |||
| AK302635.1 | - | 16 | 2282 | |||
| AK302718.1 | - | 13 | 1964 | |||
| AK302836.1 | - | 18 | 2672 | |||
| AK310356.1 | - | 13 | 1975 | |||
| AY359881.1 | - | 17 | 2395 | |||
| BC066647.1 | - | 15 | 2269 | 6166031 | ||

KIAA0586
Genomic Map