siRNA Details
| Gene Symbol: AP3D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261826.1 | NP_001248755.1 | 1, 2 | 318 | ||
| NM_003938.6 | NP_003929.4 | 1, 2 | 318 | |||
| XM_006722932.2 | XP_006722995.1 | 1, 2 | 381 | |||
| GenBank mRNA | AB208804.1 | - | 1, 2 | 302 | ||
| AF002163.1 | - | 406 | ||||
| BC005142.2 | - | 1, 2 | 146 | |||
| BC010065.2 | - | 1, 2 | 297 | 4106961 | ||
| BQ227203.1 | - | 1, 2 | 318 | |||
| U91930.1 | - | 1, 2 | 304 | |||

AP3D1
Genomic Map