siRNA Details
| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014805.3 | NP_055620.1 | 1 | 2808 | ||
| GenBank mRNA | AB018309.1 | - | 1 | 2697 | ||
| AK002069.1 | - | 1 | 1621 | |||
| AY178832.1 | - | 1 | 2808 | |||
| BC064696.1 | - | 1 | 2673 | 30345499 | ||

EPM2AIP1
Genomic Map