siRNA Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014820.4 | NP_055635.3 | 3 | 1022 | ||
| GenBank mRNA | AB018262.1 | - | 3 | 664 | ||
| AK302796.1 | - | 4 | 566 | |||
| BC003633.2 | - | 3 | 688 | 3605218 | ||
| BC052994.1 | - | 3 | 1005 | 5549394 | ||
| BC065555.1 | - | 3 | 1019 | |||

TOMM70
Genomic Map