siRNA Details
| Gene Symbol: AREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039479.1 | NP_001034568.1 | 5 | 873 | ||
| XM_005268244.3 | XP_005268301.1 | 4 | 660 | |||
| XM_006720344.2 | XP_006720407.1 | 5 | 854 | |||
| XM_011537415.2 | XP_011535717.1 | 6 | 946 | |||
| XM_011537416.2 | XP_011535718.1 | 5 | 884 | |||
| XM_011537417.2 | XP_011535719.1 | 5 | 728 | |||
| GenBank mRNA | AB002315.1 | - | 5 | 839 | ||
| AK296912.1 | - | 4 | 631 | |||
| AK304217.1 | - | 5 | 850 | |||
| BC032944.2 | - | 5 | 877 | 5267463 | ||
| BX248747.1 | - | 5 | 733 | |||

AREL1
Genomic Map