siRNA Details
| Gene Symbol: PUM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014878.4 | NP_055693.4 | 15 | 1569 | ||
| GenBank mRNA | AF490254.1 | - | 12 | 1053 | ||
| AK225953.1 | - | 15 | 1534 | |||
| AK292169.1 | - | 15 | 1512 | |||
| AK315531.1 | - | 15 | 1548 | |||
| AL832239.1 | - | 15 | 1535 | |||
| AL832245.1 | - | 15 | 1526 | |||
| BC016137.2 | - | 14 | 1473 | 3908516 | ||
| CR456957.1 | - | 12 | 1053 | |||
| D13645.1 | - | 14 | 1471 | |||

PUM3
Genomic Map