siRNA Details
| Gene Symbol: FEM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015322.4 | NP_056137.1 | 2 | 957 | ||
| XM_017021859.1 | XP_016877348.1 | 2 | 470 | |||
| GenBank mRNA | AB007856.2 | - | 2 | 342 | ||
| AF178632.1 | - | 2 | 342 | |||
| AF204883.1 | - | 2 | 342 | |||
| AK290167.1 | - | 2 | 691 | |||
| AL833264.1 | - | 1 | 1290 | |||
| BC010122.2 | - | 2 | 957 | 3840453 | ||
| BC014558.2 | - | 2 | 957 | 3840377 | ||

FEM1B
Genomic Map