siRNA Details
| Gene Symbol: RWDD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320724.1 | NP_001307653.1 | 3, 4 | 485 | ||
| NM_016940.2 | NP_058636.1 | 2, 3 | 380 | |||
| GenBank mRNA | AF212232.1 | - | 2, 3 | 341 | ||
| AK000056.1 | - | 2, 3 | 384 | |||
| AK223357.1 | - | 2, 3 | 391 | |||
| BC012546.1 | - | 2, 3 | 348 | 4291536 | ||
| BC017912.1 | - | 2, 3 | 366 | 4291966 | ||
| DB039085.1 | - | 3, 4 | 482 | |||
| Y19009.1 | - | 2, 3 | 359 | |||

RWDD2B
Genomic Map