siRNA Details
| Gene Symbol: RWDD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320724.1 | NP_001307653.1 | 6 | 1088 | ||
| NM_016940.2 | NP_058636.1 | 5 | 983 | |||
| GenBank mRNA | AF212232.1 | - | 5 | 944 | ||
| AK000056.1 | - | 5 | 987 | |||
| AK223357.1 | - | 5 | 994 | |||
| AL355726.1 | - | 1 | 137 | |||
| BC012546.1 | - | 5 | 951 | 4291536 | ||
| BC017912.1 | - | 5 | 969 | 4291966 | ||
| Y19009.1 | - | 5 | 962 | |||

RWDD2B
Genomic Map