siRNA Details
| Gene Symbol: PSTPIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321135.1 | NP_001308064.1 | 2 | 595 | ||
| NM_001321136.1 | NP_001308065.1 | 3 | 814 | |||
| NM_001321137.1 | NP_001308066.1 | 3 | 411 | |||
| NM_003978.4 | NP_003969.2 | 2 | 595 | |||
| NR_135552.1 | 2 | 595 | ||||
| XM_006720737.2 | XP_006720800.1 | 2 | 275 | |||
| XM_011522163.1 | XP_011520465.1 | 2 | 556 | |||
| XM_011522164.1 | XP_011520466.1 | 2 | 224 | |||
| XM_011522166.1 | XP_011520468.1 | 2 | 556 | |||
| XM_011522167.1 | XP_011520469.1 | 2 | 556 | |||
| XM_011522168.2 | XP_011520470.1 | 2 | 556 | |||
| XM_011522169.1 | XP_011520471.1 | 2 | 556 | |||
| GenBank mRNA | AB451310.1 | - | 2 | 106 | ||
| AB451440.1 | - | 2 | 106 | |||
| AF038602.1 | - | 2 | 106 | |||
| AF038603.1 | - | 2 | 106 | |||
| AK298733.1 | - | 2 | 296 | |||
| AK304052.1 | - | 2 | 332 | |||
| AK308462.1 | - | 3 | 411 | |||
| AK309856.1 | - | 3 | 777 | |||
| AK310072.1 | - | 3 | 422 | |||
| AK310126.1 | - | 2 | 558 | |||
| AK316220.1 | - | 2 | 306 | |||
| BC008602.1 | - | 2 | 467 | 4180398 | ||
| DB123759.1 | - | 2 | 556 | |||
| KU178458.1 | - | 2 | 106 | |||
| KU178459.1 | - | 2 | 106 | |||
| U94778.1 | - | 2 | 322 | |||

PSTPIP1
Genomic Map