siRNA Details
| Gene Symbol: MAPKAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004759.4 | NP_004750.1 | 8, 9 | 1184 | ||
| NM_032960.3 | NP_116584.2 | 8, 9 | 1184 | |||
| XM_005273353.3 | XP_005273410.1 | 8, 9 | 1292 | |||
| XM_017002810.1 | XP_016858299.1 | 8, 9 | 850 | |||
| GenBank mRNA | AK307499.1 | - | 8, 9 | 769 | ||
| BC036060.1 | - | 8, 9 | 1184 | 5296259 | ||
| BC052584.1 | - | 8, 9 | 1167 | 6267183 | ||
| X75346.1 | - | 8, 9 | 962 | |||

MAPKAPK2
Genomic Map