siRNA Details
| Gene Symbol: SH2D3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142531.1 | NP_001136003.1 | 10 | 1764 | ||
| NM_001142532.1 | NP_001136004.1 | 10 | 2013 | |||
| NM_001142533.1 | NP_001136005.1 | 9 | 1901 | |||
| NM_001142534.1 | NP_001136006.1 | 9 | 1873 | |||
| NM_001252334.1 | NP_001239263.1 | 11 | 2270 | |||
| NM_005489.3 | NP_005480.2 | 10 | 1899 | |||
| NM_170600.2 | NP_733745.1 | 11 | 2372 | |||
| XM_005251639.1 | XP_005251696.1 | 10 | 1880 | |||
| XM_011518114.1 | XP_011516416.1 | 11 | 2356 | |||
| XM_011518115.1 | XP_011516417.1 | 11 | 2316 | |||
| XM_011518117.2 | XP_011516419.1 | 9 | 2992 | |||
| XM_017014174.1 | XP_016869663.1 | 9 | 2153 | |||
| GenBank mRNA | AF124251.1 | - | 10 | 1811 | ||
| AJ420431.1 | - | 4 | 441 | |||
| AK056068.1 | - | 9 | 1901 | |||
| AK096983.1 | - | 7 | 1194 | |||
| AK128287.1 | - | 12 | 2303 | |||
| AK291393.1 | - | 9 | 1865 | |||
| AK292041.1 | - | 10 | 1827 | |||
| AK292718.1 | - | 11 | 2243 | |||
| AK294386.1 | - | 10 | 1764 | |||
| AY358089.1 | - | 10 | 1881 | |||
| AY358440.1 | - | 10 | 1800 | |||
| BC027962.1 | - | 10 | 1770 | 5223012 | ||
| BC032365.1 | - | 11 | 2247 | 4521962 | ||
| BX647905.1 | - | 11 | 2196 | |||
| CU454732.1 | - | 3 | 248 | |||
| CU455802.1 | - | 3 | 248 | |||

SH2D3C
Genomic Map