siRNA Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000349.2 | NP_000340.2 | 6 | 982 | ||
| GenBank mRNA | AF035277.1 | - | 6 | 830 | ||
| AK090513.1 | - | 4 | 1813 | |||
| AK315122.1 | - | 6 | 852 | |||
| BC010550.1 | - | 6 | 783 | 4153601 | ||
| BX647744.1 | - | 2 | 968 | |||
| CR456803.1 | - | 6 | 718 | |||
| CR541850.1 | - | 6 | 718 | |||
| KU178337.1 | - | 6 | 718 | |||
| S79669.1 | - | 6 | 870 | |||
| U17280.1 | - | 6 | 844 | |||

STAR
Genomic Map