siRNA Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000355.3 | NP_000346.2 | 6, 7 | 1191 | ||
| NM_001184726.1 | NP_001171655.1 | 6, 7 | 1110 | |||
| GenBank mRNA | AK225445.1 | - | 6, 7 | 980 | ||
| AK313711.1 | - | 6, 7 | 1081 | |||
| BC001176.1 | - | 6, 7 | 1069 | 3353271 | ||
| BC011239.1 | - | 6, 7 | 989 | 4153362 | ||
| CR456591.1 | - | 6, 7 | 981 | |||
| L02647.1 | - | 6, 7 | 1039 | |||
| L02648.1 | - | 6, 7 | 995 | |||
| M60396.1 | - | 6, 7 | 979 | |||

TCN2
Genomic Map