siRNA Details
| Gene Symbol: STIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277961.1 | NP_001264890.1 | 12 | 4057 | ||
| NM_001277962.1 | NP_001264891.1 | 13 | 3776 | |||
| NM_003156.3 | NP_003147.2 | 12 | 3739 | |||
| GenBank mRNA | BC021300.2 | - | 12 | 3731 | 4899542 | |
| CX873095.1 | - | 1 | 570 | |||
| HQ317451.1 | - | 12 | 4056 | |||
| U52426.1 | - | 12 | 3735 | |||

STIM1
Genomic Map