siRNA Details
| Gene Symbol: NEK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193533.1 | NP_001180462.1 | 15 | 3061 | ||
| NM_003157.4 | NP_003148.2 | 16 | 3328 | |||
| XM_006713310.2 | XP_006713373.1 | 15 | 3187 | |||
| GenBank mRNA | L20321.1 | - | 16 | 3303 | ||

NEK4
Genomic Map