siRNA Details
| Gene Symbol: STX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244666.1 | NP_001231595.1 | 9, 10 | 951 | ||
| NM_003164.4 | NP_003155.2 | 9, 10 | 951 | |||
| XM_011545223.2 | XP_011543525.1 | 9, 10 | 776 | |||
| XM_011545224.2 | XP_011543526.1 | 8, 9 | 667 | |||
| GenBank mRNA | AK296675.1 | - | 8, 9 | 667 | ||
| AK313497.1 | - | 9, 10 | 911 | |||
| BC002645.1 | - | 9, 10 | 901 | 3605858 | ||
| BC012137.1 | - | 9, 10 | 928 | 4560294 | ||
| BT019646.1 | - | 8, 9 | 632 | |||
| BT019647.1 | - | 8, 9 | 632 | |||
| BX537426.1 | - | 9, 10 | 949 | |||
| U26648.1 | - | 8, 9 | 658 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019829, NM_001167799, DV649852, AF232709, AK166087, and others.

STX5
Genomic Map