siRNA Details
| Gene Symbol: TGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099691.2 | NP_001093161.1 | 6 | 1709 | ||
| NM_001308158.1 | NP_001295087.1 | 6 | 1879 | |||
| NM_001308159.1 | NP_001295088.1 | 6 | 1876 | |||
| NM_003236.3 | NP_003227.1 | 6 | 1712 | |||
| GenBank mRNA | CN274505.1 | - | 1 | 459 | ||
| X70340.1 | - | 6 | 1495 | |||

TGFA
Genomic Map