siRNA Details
Gene Symbol: VSNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003385.4 | NP_003376.2 | 4 | 1590 | ||
GenBank mRNA | AF039555.1 | - | 4 | 1401 | ||
BC022012.2 | - | 4 | 1431 | 4823954 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VSNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003385.4 | NP_003376.2 | 4 | 1590 | ||
GenBank mRNA | AF039555.1 | - | 4 | 1401 | ||
BC022012.2 | - | 4 | 1431 | 4823954 |