siRNA Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143976.1 | NP_001137448.1 | 4 | 987 | ||
| NM_003390.3 | NP_003381.1 | 4 | 1127 | |||
| GenBank mRNA | AK122837.1 | - | 4 | 987 | ||
| BC051831.1 | - | 4 | 925 | 6147695 | ||
| BC070052.1 | - | 4 | 1117 | 30336707 | ||
| BX641032.1 | - | 3 | 645 | |||
| U10564.1 | - | 4 | 1127 | |||
| X62048.1 | - | 4 | 1044 | |||

WEE1
Genomic Map