siRNA Details
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003403.4 | NP_003394.1 | 5 | 1855 | ||
| GenBank mRNA | BC037308.1 | - | 5 | 1487 | 5261384 | |
| BC065366.1 | - | 5 | 1420 | 5815774 | ||
| BX647456.1 | - | 1 | 2002 | |||
| M76541.1 | - | 5 | 1449 | |||
| M77698.1 | - | 5 | 1615 | |||
| Z14077.1 | - | 5 | 1630 | |||

YY1
Genomic Map