siRNA Details
| Gene Symbol: FCGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003890.2 | NP_003881.2 | 232 | |||
| GenBank mRNA | D84239.1 | - | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FCGBP| Gene Symbol: FCGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003890.2 | NP_003881.2 | 232 | |||
| GenBank mRNA | D84239.1 | - | 232 | |||