siRNA Details
| Gene Symbol: ZBTB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143823.2 | NP_001137295.1 | 4 | 1719 | ||
| NM_001243702.1 | NP_001230631.1 | 4 | 1387 | |||
| NM_001243704.1 | NP_001230633.1 | 4 | 1378 | |||
| NM_003409.4 | NP_003400.2 | 3 | 1689 | |||
| GenBank mRNA | AK313236.1 | - | 3 | 1532 | ||
| BC057282.1 | - | 4 | 1565 | |||
| BC110519.2 | - | 2 | 1273 | 40033453 | ||
| D89859.1 | - | 2 | 1294 | |||
| Y12726.1 | - | 2 | 1261 | |||

ZBTB14
Genomic Map