siRNA Details
| Gene Symbol: ZNF143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282656.1 | NP_001269585.1 | 14 | 1764 | ||
| NM_001282657.1 | NP_001269586.1 | 13 | 1674 | |||
| NM_003442.5 | NP_003433.3 | 14 | 1767 | |||
| XM_005253121.1 | XP_005253178.1 | 14 | 1715 | |||
| XM_005253122.1 | XP_005253179.1 | 14 | 1740 | |||
| XM_005253127.1 | XP_005253184.1 | 15 | 1793 | |||
| XM_011520349.2 | XP_011518651.1 | 14 | 2486 | |||
| XM_017018252.1 | XP_016873741.1 | 13 | 1651 | |||
| XM_017018253.1 | XP_016873742.1 | 13 | 1648 | |||
| XM_017018254.1 | XP_016873743.1 | 14 | 1752 | |||
| XM_017018255.1 | XP_016873744.1 | 14 | 1842 | |||
| XM_017018256.1 | XP_016873745.1 | 13 | 1654 | |||
| GenBank mRNA | AF071771.1 | - | 11 | 1387 | ||
| AK291133.1 | - | 14 | 1720 | |||
| AK297214.1 | - | 13 | 1619 | |||
| AK313330.1 | - | 14 | 1753 | |||
| BC020219.1 | - | 14 | 1766 | 4902668 | ||

ZNF143
Genomic Map