siRNA Details
| Gene Symbol: SNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003498.5 | NP_003489.1 | 2 | 1589 | ||
| XM_017023741.1 | XP_016879230.1 | 2 | 1617 | |||
| XM_017023742.1 | XP_016879231.1 | 2 | 1515 | |||
| GenBank mRNA | AF070673.1 | - | 2 | 1572 | ||
| AK025795.1 | - | 1 | 661 | |||
| AL161976.1 | - | 2 | 1550 | |||
| BC036100.1 | - | 2 | 1567 | 5277179 | ||
| BC036443.1 | - | 2 | 1561 | 4823944 | ||

SNN
Genomic Map