siRNA Details
Gene Symbol: FZD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003507.1 | NP_003498.1 | 1 | 1986 | ||
GenBank mRNA | AB017365.1 | - | 1 | 1986 | ||
BC015915.1 | - | 1 | 1939 | 4549389 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
