siRNA Details
| Gene Symbol: HIST1H4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003545.3 | NP_003536.1 | 1 | 335 | ||
| GenBank mRNA | BC012587.1 | - | 1 | 374 | ||
| BC050615.1 | - | 1 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H4E| Gene Symbol: HIST1H4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003545.3 | NP_003536.1 | 1 | 335 | ||
| GenBank mRNA | BC012587.1 | - | 1 | 374 | ||
| BC050615.1 | - | 1 | 351 | |||