siRNA Details
| Gene Symbol: ZNF282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303481.1 | NP_001290410.1 | 6 | 1111 | ||
| NM_003575.3 | NP_003566.1 | 6 | 1111 | |||
| XM_006716151.3 | XP_006716214.1 | 6 | 1108 | |||
| GenBank mRNA | AK299280.1 | - | 6 | 1023 | ||
| AK316282.1 | - | 6 | 998 | |||
| AY927504.1 | - | 4 | 378 | |||
| AY927505.1 | - | 5 | 784 | |||
| BC041704.1 | - | 6 | 838 | |||
| BC055287.1 | - | 6 | 901 | |||
| BC073805.1 | - | 6 | 1004 | 6645861 | ||
| D30612.1 | - | 6 | 1124 | |||

ZNF282
Genomic Map