siRNA Details
| Gene Symbol: CUL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257198.1 | NP_001244127.1 | 2 | 295 | ||
| NM_003590.4 | NP_003581.1 | 2 | 612 | |||
| XM_006712800.2 | XP_006712863.2 | 2 | 285 | |||
| XM_011511994.2 | XP_011510296.1 | 2 | 283 | |||
| XM_011511995.1 | XP_011510297.1 | 2 | 197 | |||
| XM_011511996.2 | XP_011510298.1 | 1 | 135 | |||
| GenBank mRNA | AB014517.2 | - | 2 | 540 | ||
| AF062537.1 | - | 2 | 313 | |||
| AF064087.1 | - | 2 | 305 | |||
| AK291151.1 | - | 2 | 262 | |||
| AK308496.1 | - | 2 | 295 | |||
| AY337761.1 | - | 2 | 758 | |||
| BC039598.1 | - | 2 | 557 | 5784147 | ||
| BC092409.1 | - | 2 | 557 | 6168545 | ||
| BQ688156.1 | - | 612 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_016716, BC027304, AK129174, AF129738.

CUL3
Genomic Map