siRNA Details
| Gene Symbol: IRS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003604.2 | NP_003595.1 | 1 | 722 | ||
| XM_005262220.2 | XP_005262277.1 | 1 | 929 | |||
| XM_006724713.3 | XP_006724776.1 | 1 | 929 | |||
| XM_011531061.1 | XP_011529363.1 | 1 | 929 | |||
| GenBank mRNA | AF007567.1 | - | 1 | 767 | ||

IRS4
Genomic Map