siRNA Details
| Gene Symbol: MBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003927.4 | NP_003918.1 | 1 | 723 | ||
| NM_015832.4 | NP_056647.1 | 1 | 723 | |||
| GenBank mRNA | AF072242.1 | - | 1 | 723 | ||
| AF072246.1 | - | 1 | 26 | |||
| BC032638.1 | - | 1 | 564 | 5496721 | ||
| BQ228712.1 | - | 442 | ||||

MBD2
Genomic Map