siRNA Details
| Gene Symbol: NIPSNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202502.1 | NP_001189431.1 | 2 | 165 | ||
| NM_003634.3 | NP_003625.2 | 2 | 466 | |||
| GenBank mRNA | AJ001258.1 | - | 2 | 465 | ||
| AK098367.1 | - | 2 | 220 | |||
| AK290953.1 | - | 2 | 219 | |||
| AK298816.1 | - | 2 | 167 | |||
| AK314407.1 | - | 2 | 237 | |||
| BC002371.2 | - | 2 | 216 | 2822369 | ||
| BC006473.1 | - | 2 | 216 | 2822369 | ||
| BC018869.2 | - | 1 | 108 | |||
| DA133702.1 | - | 2 | 284 | |||

NIPSNAP1
Genomic Map