siRNA Details
| Gene Symbol: PRKRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139517.1 | NP_001132989.1 | 1 | 392 | ||
| NM_001139518.1 | NP_001132990.1 | 2 | 113 | |||
| NM_001316362.1 | NP_001303291.1 | 2 | 321 | |||
| NM_003690.4 | NP_003681.1 | 2 | 312 | |||
| XM_011512063.1 | XP_011510365.1 | 2 | 194 | |||
| XM_011512066.1 | XP_011510368.1 | 2 | 358 | |||
| XM_017005159.1 | XP_016860648.1 | 2 | 194 | |||
| GenBank mRNA | AF072860.1 | - | 2 | 207 | ||
| AF083033.1 | - | 2 | 203 | |||
| AK223107.1 | - | 2 | 211 | |||
| AK290601.1 | - | 2 | 238 | |||
| AK296019.1 | - | 2 | 232 | |||
| AL136615.1 | - | 2 | 236 | |||
| AL833867.1 | - | 1 | 392 | |||
| AY251164.1 | - | 2 | 130 | |||
| BC009470.1 | - | 2 | 218 | 3503747 | ||
| BG827125.1 | - | 2 | 313 | |||
| BT007243.1 | - | 2 | 111 | |||
| CR533525.1 | - | 2 | 111 | |||
| HY049565.1 | - | 2 | 321 | |||

PRKRA
Genomic Map