siRNA Details
| Gene Symbol: UNC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003728.3 | NP_003719.3 | 2 | 673 | ||
| XM_005263321.3 | XP_005263378.1 | 2 | 673 | |||
| XM_017008789.1 | XP_016864278.1 | 3 | 7090 | |||
| XM_017008790.1 | XP_016864279.1 | 2 | 318 | |||
| GenBank mRNA | AF055634.1 | - | 2 | 474 | ||
| AK290500.1 | - | 2 | 484 | |||
| AK308421.1 | - | 2 | 436 | |||
| BC041156.1 | - | 2 | 669 | 5208108 | ||

UNC5C
Genomic Map