siRNA Details
| Gene Symbol: TBCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297738.1 | NP_001284667.1 | 3 | 2171 | ||
| NM_001297740.1 | NP_001284669.1 | 3 | 317 | |||
| NM_004607.2 | NP_004598.1 | 4 | 404 | |||
| GenBank mRNA | AF038952.1 | - | 4 | 349 | ||
| AK312207.1 | - | 4 | 372 | |||
| BC018210.1 | - | 4 | 389 | 3861138 | ||
| BC105285.1 | - | 4 | 340 | 6702573 | ||
| BT007354.1 | - | 4 | 300 | |||
| CR542170.1 | - | 4 | 300 | |||
| CR542186.1 | - | 4 | 300 | |||
| DB538817.1 | - | 2 | 167 | |||
| HY015414.1 | - | 3 | 294 | |||

TBCA
Genomic Map