siRNA Details
| Gene Symbol: USP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004651.3 | NP_004642.2 | 12 | 1664 | ||
| XM_005272674.3 | XP_005272731.1 | 12 | 1501 | |||
| XM_011543988.1 | XP_011542290.1 | 12 | 1458 | |||
| GenBank mRNA | AB073597.1 | - | 12 | 1566 | ||
| AK055588.1 | - | 11 | 1721 | |||
| AK098394.1 | - | 12 | 1568 | |||
| AK294635.1 | - | 12 | 1410 | |||
| BC000350.2 | - | 12 | 1538 | |||
| BC063668.1 | - | 12 | 1546 | |||
| BC140849.1 | - | 12 | 1713 | 9021717 | ||
| U44839.1 | - | 12 | 1524 | |||

USP11
Genomic Map