siRNA Details
| Gene Symbol: FXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013438.2 | NP_001013456.1 | 3 | 402 | ||
| NM_001013439.2 | NP_001013457.1 | 4 | 746 | |||
| NM_005087.3 | NP_005078.2 | 3 | 402 | |||
| XM_005247813.3 | XP_005247870.2 | 3 | 177 | |||
| XM_005247814.3 | XP_005247871.2 | 3 | 177 | |||
| XM_005247815.3 | XP_005247872.2 | 3 | 177 | |||
| XM_005247816.3 | XP_005247873.2 | 3 | 177 | |||
| XM_006713775.3 | XP_006713838.1 | 4 | 520 | |||
| XM_017007298.1 | XP_016862787.1 | 4 | 520 | |||
| XM_017007299.1 | XP_016862788.1 | 4 | 520 | |||
| XM_017007300.1 | XP_016862789.1 | 4 | 520 | |||
| XM_017007301.1 | XP_016862790.1 | 4 | 520 | |||
| GenBank mRNA | AK225334.1 | - | 3 | 178 | ||
| AK292633.1 | - | 3 | 178 | |||
| AK294603.1 | - | 2 | 124 | |||
| AK302197.1 | - | 3 | 181 | |||
| AU280585.1 | - | 4 | 521 | |||
| AY341428.1 | - | 3 | 453 | |||
| BC028983.1 | - | 3 | 172 | 5122574 | ||
| BX956500.1 | - | 3 | 188 | |||
| DC352767.1 | - | 3 | 402 | |||
| KR259877.1 | - | 3 | 162 | |||
| KR259878.1 | - | 3 | 162 | |||
| KR259879.1 | - | 3 | 162 | |||
| KR259880.1 | - | 4 | 506 | |||
| U25165.1 | - | 3 | 174 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001113188, NM_008053, NM_001113189, BC019139, X90875, and others.

FXR1
Genomic Map