siRNA Details
| Gene Symbol: FXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013438.2 | NP_001013456.1 | 15 | 1681 | ||
| NM_001013439.2 | NP_001013457.1 | 16 | 2025 | |||
| NM_005087.3 | NP_005078.2 | 15 | 1681 | |||
| XM_005247813.3 | XP_005247870.2 | 14 | 1543 | |||
| XM_005247814.3 | XP_005247871.2 | 14 | 1543 | |||
| XM_005247815.3 | XP_005247872.2 | 15 | 1456 | |||
| XM_005247816.3 | XP_005247873.2 | 14 | 1543 | |||
| XM_006713775.3 | XP_006713838.1 | 15 | 1886 | |||
| XM_017007298.1 | XP_016862787.1 | 15 | 1886 | |||
| XM_017007299.1 | XP_016862788.1 | 16 | 1799 | |||
| XM_017007300.1 | XP_016862789.1 | 15 | 1886 | |||
| XM_017007301.1 | XP_016862790.1 | 16 | 1799 | |||
| GenBank mRNA | AK225334.1 | - | 15 | 1457 | ||
| AK292633.1 | - | 15 | 1457 | |||
| AK294603.1 | - | 14 | 1403 | |||
| AK297254.1 | - | 13 | 1309 | |||
| AK297331.1 | - | 13 | 1449 | |||
| AK302197.1 | - | 15 | 1460 | |||
| AY341428.1 | - | 15 | 1732 | |||
| BC028983.1 | - | 15 | 1451 | 5122574 | ||
| KR259877.1 | - | 15 | 1441 | |||
| KR259878.1 | - | 14 | 1528 | |||
| KR259879.1 | - | 14 | 1528 | |||
| KR259880.1 | - | 16 | 1753 | |||
| U25165.1 | - | 15 | 1453 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001113188, NM_008053, NM_001113189, AK049231, BC019139, and others.

FXR1
Genomic Map