siRNA Details
| Gene Symbol: NELFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005663.4 | NP_005654.3 | 11 | 1850 | ||
| XM_017008589.1 | XP_016864078.1 | 12 | 2671 | |||
| GenBank mRNA | AB044549.1 | - | 10 | 1637 | ||
| AF101434.1 | - | 11 | 1579 | |||
| AF131751.1 | - | 7 | 1100 | |||
| AK001304.1 | - | 11 | 1637 | |||
| AK094040.1 | - | 10 | 1410 | |||
| AK095737.1 | - | 10 | 1893 | |||
| AK126056.1 | - | 11 | 2725 | |||
| AK222853.1 | - | 11 | 1587 | |||
| BC002764.2 | - | 11 | 1590 | 3630594 | ||

NELFA
Genomic Map