siRNA Details
| Gene Symbol: STK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005990.3 | NP_005981.3 | 9, 10 | 1862 | ||
| XM_017009788.1 | XP_016865277.1 | 8, 9 | 1416 | |||
| GenBank mRNA | AB015718.1 | - | 9, 10 | 1612 | ||
| AK022960.1 | - | 1, 2 | 435 | |||
| AK307943.1 | - | 9, 10 | 1396 | |||
| AK313350.1 | - | 10, 11 | 1617 | |||
| BC070077.1 | - | 9, 10 | 1776 | 5267449 | ||

STK10
Genomic Map