siRNA Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003818.3 | NP_003809.1 | 13 | 1805 | ||
| XM_006723660.1 | XP_006723723.1 | 13 | 1816 | |||
| XM_011529390.1 | XP_011527692.1 | 14 | 2128 | |||
| GenBank mRNA | AF069532.1 | - | 12 | 1401 | ||
| AK001414.1 | - | 13 | 1525 | |||
| AK027695.1 | - | 13 | 1513 | |||
| AK095430.1 | - | 10 | 1728 | |||
| Y16521.1 | - | 13 | 1730 | |||

CDS2
Genomic Map