siRNA Details
| Gene Symbol: MAPK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139021.2 | NP_620590.2 | 14 | 1548 | ||
| XM_011516925.2 | XP_011515227.1 | 14 | 2386 | |||
| XM_011516926.2 | XP_011515228.1 | 13 | 1831 | |||
| XM_011516927.2 | XP_011515229.1 | 13 | 2523 | |||
| XM_017013218.1 | XP_016868707.1 | 12 | 2487 | |||
| GenBank mRNA | AY065978.1 | - | 14 | 1540 | ||
| BC028034.1 | - | 12 | 1905 | 5242481 | ||

MAPK15
Genomic Map