siRNA Details
| Gene Symbol: TYRO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006293.3 | NP_006284.2 | 4 | 746 | ||
| XM_017022542.1 | XP_016878031.1 | 4 | 711 | |||
| XM_017022543.1 | XP_016878032.1 | 4 | 547 | |||
| GenBank mRNA | AB209431.1 | - | 4 | 775 | ||
| BC034960.1 | - | 4 | 528 | |||
| BC049368.1 | - | 4 | 520 | 4821587 | ||
| BC051756.1 | - | 4 | 525 | 6095596 | ||
| D17517.1 | - | 4 | 734 | |||
| D50479.1 | - | 4 | 506 | |||
| U05682.1 | - | 4 | 516 | |||

TYRO3
Genomic Map