siRNA Details
| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003583.3 | NP_003574.1 | 2 | 2694 | ||
| NM_006482.2 | NP_006473.2 | 3 | 2843 | |||
| XM_017020032.1 | XP_016875521.1 | 2 | 2986 | |||
| GenBank mRNA | AK025018.1 | - | 1 | 184 | ||
| CN303286.1 | - | 1 | 251 | |||
| Y13493.1 | - | 3 | 2601 | |||

DYRK2
Genomic Map