siRNA Details
| Gene Symbol: NAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003827.3 | NP_003818.2 | 3, 4 | 623 | ||
| NR_038456.1 | 3, 4 | 623 | ||||
| NR_038457.1 | 1, 2 | 426 | ||||
| XM_011527436.1 | XP_011525738.1 | 3, 4 | 297 | |||
| XM_011527437.1 | XP_011525739.1 | 2, 3 | 376 | |||
| GenBank mRNA | AK098401.1 | - | 3, 4 | 456 | ||
| AK126519.1 | - | 1 | 1510 | |||
| AK292244.1 | - | 3, 4 | 387 | |||
| AK308633.1 | - | 4, 5 | 513 | |||
| AK310114.1 | - | 1, 2 | 190 | |||
| BC001165.1 | - | 3, 4 | 395 | 3533668 | ||
| BC007432.2 | - | 3, 4 | 386 | 2962847 | ||
| BC028234.1 | - | 3, 4 | 441 | 5245770 | ||
| BC091511.1 | - | 3, 4 | 402 | 30512911 | ||
| BQ948081.1 | - | 3, 4 | 605 | |||
| BT019568.1 | - | 3, 4 | 305 | |||
| CX873074.1 | - | 1, 2 | 168 | |||
| U39412.1 | - | 3, 4 | 372 | |||

NAPA
Genomic Map