siRNA Details
| Gene Symbol: GLRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042543.2 | NP_001036008.1 | 7 | 1422 | ||
| NM_006529.3 | NP_006520.2 | 7 | 1422 | |||
| XM_011532267.2 | XP_011530569.1 | 7 | 1675 | |||
| XM_011532268.2 | XP_011530570.1 | 3 | 668 | |||
| XM_017008630.1 | XP_016864119.1 | 8 | 1446 | |||
| GenBank mRNA | AF018157.1 | - | 1 | 185 | ||
| BC036086.1 | - | 7 | 1421 | 5286731 | ||
| CV381684.1 | - | 1 | 175 | |||
| U93917.1 | - | 7 | 1338 | |||

GLRA3
Genomic Map