siRNA Details
| Gene Symbol: IFRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006764.4 | NP_006755.4 | 11 | 1490 | ||
| GenBank mRNA | AK074763.1 | - | 11 | 1307 | ||
| AK097765.1 | - | 1 | 1405 | |||
| BC001327.1 | - | 11 | 1317 | 3456988 | ||
| BC001676.2 | - | 11 | 1302 | 2963821 | ||
| BC007265.1 | - | 11 | 1325 | 3138466 | ||
| BC007437.2 | - | 11 | 1350 | 2961581 | ||
| BT019999.1 | - | 11 | 1169 | |||
| U09585.1 | - | 11 | 1167 | |||
| Y12395.1 | - | 14 | 1667 | |||

IFRD2
Genomic Map